Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sek-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_076921.7 | NP_509322.2 | 4 - 5 | 694 | 66 | |
GenBank mRNA | AB024087.1 | - | 4 - 5 | 704 | 66 | |
AB060731.2 | - | 4 - 5 | 704 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |