Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tnt-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001373379.1 | NP_001360658.1 | 7 - 8 | 720 | 68 | |
NM_001373380.2 | NP_001359846.1 | 7 - 8 | 672 | 68 | ||
NM_001373381.2 | NP_001359848.1 | 6 - 7 | 3347 | 68 | ||
NM_001373382.2 | NP_001360657.1 | 6 - 7 | 3346 | 68 | ||
NM_001373383.1 | NP_001359847.1 | 6 - 7 | 3347 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |