Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sdpn-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001381034.1 | NP_001367622.1 | 4 - 5 | 421 | 109 | |
NM_001381035.1 | NP_001367621.1 | 4 - 5 | 555 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Caenorhabditis elegans Probe spans exons |