Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sdpn-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001381034.1 | NP_001367622.1 | 8 - 9 | 909 | 92 | |
NM_001381035.1 | NP_001367621.1 | 8 - 9 | 1043 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |