Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mec-10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_077037.7 | NP_509438.1 | 13 - 14 | 1701 | 103 | |
GenBank mRNA | L25312.1 | - | 13 - 14 | 1643 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |