Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rig-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001375088.2 | NP_001362001.1 | 2 - 3 | 148 | 117 | |
NM_001383616.1 | 1 - 2 | 106 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |