Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sec-15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_077095.10 | NP_509496.3 | 11 - 12 | 2349 | 85 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |