Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sec-15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_077095.10 | NP_509496.3 | 8 - 9 | 1480 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |