Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ajm-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001330906.2 | NP_001317734.1 | 19 - 20 | 4083 | 80 | |
NM_001330908.3 | NP_001317889.1 | 13 - 14 | 3263 | 80 | ||
NM_001330909.3 | NP_001317890.1 | 13 - 14 | 3038 | 80 | ||
NM_001330910.3 | NP_001317891.1 | 11 - 12 | 2906 | 80 | ||
NM_001330911.3 | NP_001317888.1 | 10 - 11 | 2792 | 80 | ||
NM_001330912.3 | NP_001317897.1 | 9 - 10 | 2476 | 80 | ||
NM_001330913.3 | NP_001317898.1 | 8 - 9 | 2231 | 80 | ||
NM_001330914.3 | NP_001317899.1 | 7 - 8 | 2159 | 80 | ||
NM_001330915.3 | NP_001317736.1 | 5 - 6 | 1862 | 80 | ||
NM_001381045.1 | NP_001368606.1 | 19 - 20 | 4046 | 80 | ||
NM_001381048.1 | NP_001367381.1 | 19 - 20 | 4083 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |