Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gpa-12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_077156.6 | NP_509557.1 | 7 - 8 | 775 | 61 | |
GenBank mRNA | AY008134.1 | - | 6 - 7 | 703 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |