Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: oga-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001129711.3 | NP_001123183.1 | 8 - 9 | 1313 | 88 | |
NM_001129712.4 | NP_001123184.1 | 7 - 8 | 1363 | 88 | ||
GenBank mRNA | DQ407522.1 | - | 8 - 9 | 1263 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |