Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gap-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001129726.4 | NP_001123198.1 | 3 - 4 | 798 | 81 | |
NM_001307914.4 | NP_001294843.1 | 7 - 8 | 1264 | 81 | ||
NM_001307916.4 | NP_001294845.1 | 12 - 13 | 2041 | 81 | ||
NM_001383623.1 | 10 - 11 | 1645 | 81 | |||
NM_077193.6 | NP_509594.3 | 12 - 13 | 2042 | 81 | ||
NM_077194.7 | NP_509595.1 | 13 - 14 | 2104 | 81 | ||
NM_077195.7 | NP_509596.1 | 11 - 12 | 2692 | 81 | ||
NM_077196.6 | NP_509597.1 | 10 - 11 | 1831 | 81 | ||
NM_077197.6 | NP_509598.1 | 9 - 10 | 1581 | 81 | ||
NM_077198.7 | NP_509599.1 | 7 - 8 | 2161 | 81 | ||
NM_171753.7 | NP_741873.1 | 10 - 11 | 2064 | 81 | ||
NM_171754.6 | NP_741874.1 | 3 - 4 | 787 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |