Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gap-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001129726.4 | NP_001123198.1 | 5 - 6 | 1395 | 81 | |
NM_001307914.4 | NP_001294843.1 | 9 - 10 | 1861 | 81 | ||
NM_001307916.4 | NP_001294845.1 | 14 - 15 | 2638 | 81 | ||
NM_001383623.1 | 12 - 13 | 2242 | 81 | |||
NM_077193.6 | NP_509594.3 | 14 - 15 | 2639 | 81 | ||
NM_077194.7 | NP_509595.1 | 15 - 16 | 2701 | 81 | ||
NM_077195.7 | NP_509596.1 | 13 - 14 | 3289 | 81 | ||
NM_077196.6 | NP_509597.1 | 12 - 13 | 2428 | 81 | ||
NM_077197.6 | NP_509598.1 | 11 - 12 | 2178 | 81 | ||
NM_077198.7 | NP_509599.1 | 9 - 10 | 2758 | 81 | ||
NM_171753.7 | NP_741873.1 | 12 - 13 | 2661 | 81 | ||
NM_171754.6 | NP_741874.1 | 5 - 6 | 1384 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |