Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gap-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001383623.1 | 3 - 4 | 343 | 70 | ||
NM_077194.7 | NP_509595.1 | 6 - 7 | 802 | 70 | ||
NM_077195.7 | NP_509596.1 | 4 - 5 | 1390 | 70 | ||
NM_077196.6 | NP_509597.1 | 3 - 4 | 529 | 70 | ||
NM_171753.7 | NP_741873.1 | 3 - 4 | 762 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |