Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mgl-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001129721.4 | NP_001123193.2 | 4 - 5 | 287 | 96 | |
NM_001313458.3 | NP_001300387.1 | 5 - 6 | 329 | 96 | ||
NM_001313459.3 | NP_001300388.1 | 1 - 2 | 62 | 96 | ||
NM_077273.4 | NP_509674.2 | 5 - 6 | 546 | 96 |
Set Membership: |
Amplicon length between 86 and 100 G-protein coupled receptor Detect Genomic DNA Caenorhabditis elegans Probe spans exons |