Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: moc-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_077299.6 | NP_509700.2 | 5 - 6 | 1417 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |