Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sex-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029491.5 | NP_001024662.1 | 5 - 6 | 854 | 115 | |
GenBank mRNA | U13074.1 | - | 5 - 6 | 875 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |