Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: acl-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047817.4 | NP_001041282.1 | 6 - 7 | 900 | 67 | |
NM_001047818.4 | NP_001041283.1 | 5 - 6 | 728 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |