Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rme-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001373469.3 | NP_001359519.1 | 4 | 452 | 96 | |
NM_077338.5 | NP_509739.1 | 5 - 6 | 643 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |