Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rme-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001373469.3 | NP_001359519.1 | 10 | 1440 | 66 | |
NM_077338.5 | NP_509739.1 | 11 - 12 | 1631 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |