Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mig-15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029800.3 | NP_001024971.1 | 2 - 3 | 290 | 123 | |
NM_001029801.5 | NP_001024972.1 | 2 - 3 | 290 | 123 | ||
NM_001029802.5 | NP_001024973.1 | 2 - 3 | 214 | 123 | ||
NM_001029803.6 | NP_001024974.1 | 2 - 3 | 297 | 123 | ||
GenBank mRNA | AF087131.1 | - | 2 - 3 | 312 | 123 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |