Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mig-15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029802.5 | NP_001024973.1 | 10 - 11 | 2055 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |