Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: vab-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029399.4 | NP_001024570.1 | 4 - 5 | 531 | 67 | |
GenBank mRNA | U31537.1 | - | 4 - 5 | 485 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |