Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: syd-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_077367.9 | NP_509768.1 | 16 - 17 | 2547 | 94 | |
GenBank mRNA | AF170122.1 | - | 15 - 16 | 2474 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |