Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: abl-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001381086.2 | NP_001368776.1 | 14 - 15 | 3409 | 87 | |
NM_001381087.2 | NP_001368775.1 | 16 - 17 | 3499 | 87 | ||
NM_077376.10 | NP_509777.2 | 15 - 16 | 3482 | 87 | ||
NM_077377.6 | NP_509778.2 | 16 - 17 | 3502 | 87 | ||
NM_077378.5 | NP_509779.2 | 12 - 13 | 3559 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |