Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: plc-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029446.3 | NP_001024617.1 | 9 - 10 | 1732 | 64 | |
NM_001029448.6 | NP_001024619.1 | 9 - 10 | 1694 | 64 | ||
NM_001129669.4 | NP_001123141.2 | 42 - 43 | 6572 | 64 | ||
NM_001136455.4 | NP_001129927.1 | 9 - 10 | 1916 | 64 | ||
NM_001373489.2 | NP_001360637.1 | 42 - 43 | 6551 | 64 | ||
NM_001373491.2 | NP_001360639.1 | 42 - 43 | 6581 | 64 | ||
NM_001373492.2 | NP_001360635.1 | 42 - 43 | 6602 | 64 | ||
NM_001373493.2 | NP_001360636.1 | 43 - 44 | 6722 | 64 | ||
NM_001373494.2 | NP_001359720.1 | 43 - 44 | 6743 | 64 | ||
NM_001373495.2 | NP_001360638.1 | 43 - 44 | 6752 | 64 | ||
NM_077404.6 | NP_509805.3 | 9 - 10 | 1694 | 64 | ||
GenBank mRNA | AF044576.1 | - | 9 - 10 | 1754 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |