Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: plc-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001129669.4 | NP_001123141.2 | 26 - 27 | 3907 | 79 | |
NM_001373489.2 | NP_001360637.1 | 26 - 27 | 3886 | 79 | ||
NM_001373491.2 | NP_001360639.1 | 26 - 27 | 3916 | 79 | ||
NM_001373492.2 | NP_001360635.1 | 26 - 27 | 3937 | 79 | ||
NM_001373493.2 | NP_001360636.1 | 27 - 28 | 4057 | 79 | ||
NM_001373494.2 | NP_001359720.1 | 27 - 28 | 4078 | 79 | ||
NM_001373495.2 | NP_001360638.1 | 27 - 28 | 4087 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |