Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pbo-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_077429.8 | NP_509830.2 | 8 - 9 | 1404 | 72 | |
GenBank mRNA | AF497831.1 | - | 8 - 9 | 1426 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |