Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lge-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_077432.6 | NP_509833.3 | 8 - 9 | 1517 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |