Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: egl-15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001313426.3 | NP_001300355.1 | 18 | 2991 | 96 | |
NM_001313427.4 | NP_001300356.1 | 18 | 2991 | 96 | ||
NM_001313435.5 | NP_001300364.1 | 18 | 2827 | 96 | ||
NM_001373496.2 | NP_001360406.1 | 18 | 2990 | 96 | ||
NM_001381095.1 | NP_001368580.1 | 17 - 18 | 2794 | 96 | ||
NM_001381096.1 | NP_001368579.1 | 17 - 18 | 2794 | 96 | ||
NM_001381098.1 | NP_001367004.1 | 17 - 18 | 2794 | 96 | ||
NM_001383640.1 | 17 - 18 | 2991 | 96 | |||
GenBank mRNA | U39761.1 | - | 17 - 18 | 2959 | 96 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |