Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: egl-15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001313426.3 | NP_001300355.1 | 6 | 777 | 114 | |
NM_001313427.4 | NP_001300356.1 | 6 | 777 | 114 | ||
NM_001373496.2 | NP_001360406.1 | 6 | 776 | 114 | ||
NM_001383640.1 | 5 - 6 | 777 | 114 | |||
GenBank mRNA | U39761.1 | - | 5 - 6 | 745 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Amplicon spans exons and probe does not span exons |