Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sulp-7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038479.2 | NP_001033568.1 | 4 - 5 | 400 | 71 | |
NM_001038480.2 | NP_001033569.1 | 4 - 5 | 460 | 71 | ||
NM_001038481.4 | NP_001033570.1 | 6 - 7 | 732 | 71 | ||
NM_001038482.4 | NP_001033571.1 | 6 - 7 | 732 | 71 | ||
GenBank mRNA | AY887913.1 | - | 4 - 5 | 448 | 71 | |
AY887914.1 | - | 4 - 5 | 505 | 71 | ||
AY887915.1 | - | 6 - 7 | 754 | 71 | ||
AY887916.1 | - | 6 - 7 | 754 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |