Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hst-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_077470.6 | NP_509871.2 | 2 - 3 | 309 | 144 | |
GenBank mRNA | AY552606.1 | - | 2 - 3 | 242 | 144 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |