Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: inx-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_077484.5 | NP_509885.1 | 1 - 2 | 536 | 142 | |
GenBank mRNA | AF304127.1 | - | 1 - 2 | 530 | 142 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |