Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: npr-9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001375132.1 | NP_001361821.1 | 4 - 5 | 589 | 71 |
Set Membership: |
Amplicon length between 71 and 85 G-protein coupled receptor Detect Genomic DNA Caenorhabditis elegans Probe spans exons |