Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gcy-9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_077496.5 | NP_509897.3 | 22 - 23 | 3316 | 72 | |
GenBank mRNA | HQ636455.1 | - | 21 - 22 | 2925 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |