Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pek-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_077511.8 | NP_509912.1 | 3 - 4 | 887 | 103 | |
GenBank mRNA | AF193341.1 | - | 3 - 4 | 811 | 103 | |
AF435953.1 | - | 3 - 4 | 811 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |