Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pek-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_077511.8 | NP_509912.1 | 8 - 9 | 1998 | 68 | |
GenBank mRNA | AF193341.1 | - | 8 - 9 | 1922 | 68 | |
AF435953.1 | - | 8 - 9 | 1922 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |