Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lev-8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_077531.7 | NP_509932.2 | 10 - 11 | 1300 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |