Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ftt-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001350351.5 | NP_001337315.1 | 2 | 89 | 51 | |
NM_077538.8 | NP_509939.1 | 1 - 2 | 121 | 51 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |