Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sto-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_077540.2 | NP_509941.1 | 3 - 4 | 439 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |