Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ttx-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001270239.2 | NP_001257168.1 | 8 - 9 | 1012 | 72 | |
NM_001270240.3 | NP_001257169.1 | 7 - 8 | 928 | 72 | ||
GenBank mRNA | U72211.1 | - | 8 - 9 | 1012 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |