Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cab-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_077635.6 | NP_510036.1 | 7 - 8 | 1181 | 78 | |
GenBank mRNA | AF293976.1 | - | 5 - 6 | 994 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |