Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srd-47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_077665.1 | NP_510066.1 | 5 - 6 | 779 | 100 |
Set Membership: |
Amplicon length between 86 and 100 G-protein coupled receptor 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |