Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: zig-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_077668.5 | NP_510069.1 | 2 - 3 | 451 | 79 | |
GenBank mRNA | AF456249.1 | - | 2 - 3 | 396 | 79 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |