Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nekl-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_077679.8 | NP_510080.2 | 5 - 6 | 587 | 95 | |
GenBank mRNA | DQ645955.1 | - | 5 - 6 | 603 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |