Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: atf-6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_077693.6 | NP_510094.1 | 8 - 9 | 1467 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |