Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: adt-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001375156.2 | NP_001361951.1 | 12 - 13 | 1893 | 80 | |
NM_077715.5 | NP_510116.2 | 12 - 13 | 1902 | 80 | ||
GenBank mRNA | AB066246.1 | - | 12 - 13 | 1902 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |