Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: abf-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_077735.5 | NP_510136.1 | 1 - 2 | 288 | 67 | |
GenBank mRNA | AB086409.1 | - | 1 - 2 | 163 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |