Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pqn-18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001270274.4 | NP_001257203.1 | 2 - 3 | 282 | 79 | |
NM_001270275.4 | NP_001257204.1 | 1 - 2 | 195 | 79 | ||
NM_001381127.1 | NP_001366740.1 | 4 - 5 | 902 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |